Methylphenylsulfid for synthesis
Leverandør: Novabiochem (Part of Merck)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Methyl phenyl sulphide, Phenyl methyl sulphide, (Methylthio)benzene
M-Clarity™ quality level = MQ200
8.20825.0100 8.20825.2500 8.20825.0500 8.20825.0100 8.20825.2500
8.20825.0100EA 0 DKK
8.20825.0100 NOVA8.20825.2500 8.20825.0500 MERC8.20825.0100 8.20825.2500
Methylphenylsulfid for synthesis
Methylphenylsulfid
Formel: C₇H₈S
Molekylvægt: 124.21 g/mol
Kogepunkt: 188
Smeltepunkt: –15 °C
Massefylde: 1.05 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008559
CAS-nummer: 100-68-5
EINECS: 202-878-2
UN nummer: 3082
ADR: 9 III