4-Fluor-2-methoxybenzoesyre ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H26179.03 H26179.06
H26179.03EA 316 DKK
H26179.03 H26179.06
4-Fluor-2-methoxybenzoesyre ≥99%
4-Fluor-2-methoxybenzoesyre

Purity: 99% CAS No.: 395-82-4 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 134-136° MDL No.: MFCD00673010

Formel: C₈H₇FO₃
Smeltepunkt: 134…136 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00673010
CAS-nummer: 395-82-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR