4-fluorbenzamid 98

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L06831.14 L06831.06 L06831.22
L06831.14EA 1210 DKK
L06831.14 L06831.06 L06831.22
4-fluorbenzamid 98
4-fluorbenzamid

Purity: 98% CAS No.: 824-75-9 Molecular Formula: C7H6FNO Formula Weight: 139.13 Melting Point: 154-157° Beilstein Reference: 1859744

Formel: C₇H₆FNO
Molekylvægt: 139.13 g/mol
Kogepunkt: 287
Smeltepunkt: 154…157 °C
Massefylde: 1.43 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007992
CAS-nummer: 824-75-9
EINECS: 212-535-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR