VWR®, Watch Glasses
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-2181EA 401 DKK
216-2181 216-2182 216-2183 216-2184 216-2185 216-2186 216-2187 216-2188
VWR®, Watch Glasses
Urglas
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable and sterilisable
These watch glasses are concave shaped, and can be used to evaporate liquids, hold solids while being weighed, or as vessel cover.