Kaliumhydrogenphthalat, EMSURE® Reag. Ph. Eur. for analysis, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt

M-Clarity™ quality level = MQ300

1.04874.0250 1.04874.1000 1.04874.9050
1.04874.0250EA 690 DKK
1.04874.0250 1.04874.1000 1.04874.9050
Kaliumhydrogenphthalat, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Kaliumhydrogenphthalat
Formel: HOOCC₆H₄COOK
Molekylvægt: 204.22 g/mol
Smeltepunkt: 295…300 °C
Massefylde: 1.6362 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013070
CAS-nummer: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 80 g/l (20 °C)
Melting Point 295 - 300 °C
Molar Mass 204.22 g/mol
Bulk Density 900 kg/m3
Density 1.636 g/cm3
pH 4.0 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR