Dimethylphthalat ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Phthalic acid dimethyl ester

P0302-25G P0302-500G
TCIAP0302-25GEA 132 DKK
TCIAP0302-25G TCIAP0302-500G
Dimethylphthalat ≥99.0% (by GC)
Dimethylphthalat
Formel: C₁₀H₁₀O₄
Molekylvægt: 194.19 g/mol
Kogepunkt: 284°C
Smeltepunkt: 6
Flammepunkt: 156°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008425
CAS-nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 14,03255
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5252432eb117f168062cb5e35ebec20f1ef7431&vid=a5252432eb117f168062cb5e35ebec20f1ef7431&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR