Ethyl 3-hydroxybenzoat ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-Hydroxybenzoic acid ethyl ester, Ethyl 3-hydroxybenzoate

A15343.14 A15343.22 A15343.36
A15343.14EA 658 DKK
A15343.14 A15343.22 A15343.36
Ethyl 3-hydroxybenzoat ≥99%
Ethyl 3-hydroxybenzoat

Purity: 99% CAS No.: 7781-98-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 72-76° Boiling Point: 187-188°/31mm MDL No.: MFCD00002296

Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
Kogepunkt: 187…188 °C (31 mmHg)
Smeltepunkt: 72…76 °C
Massefylde: 1.068 g/cm³ (131 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002296
CAS-nummer: 7781-98-8
EINECS: 231-951-1

Bestil nu

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR