Ethylsalicylat 99+%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Ethyl 2-hydroxybenzoate
150490010 150492500 150490250
ACRO150490010EA 714 DKK
ACRO150490010 ACRO150492500 ACRO150490250
Ethylsalicylat 99+%
Ethylsalicylat
Formel: C₉H₁₀O₃
Molekylvægt: 166.18 g/mol
Kogepunkt: 234 °C (760 mmHg)
Smeltepunkt: 2…3 °C
Massefylde: 1.128 g/cm³
Flammepunkt: 107 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002215
CAS-nummer: 118-61-6
EINECS: 204-265-5
Merck Index: 13,03881
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5250 (20°C, 589 nm)
GC ≥99.0 %
Specific gravity (25/25°C) 1.126 to 1.13
Acid value ≤1 mg KOH/g