Benzoesyrephenylamid ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

A11086.22 A11086.36
A11086.22EA 521 DKK
A11086.22 A11086.36
Benzoesyrephenylamid ≥98%
Benzoesyrephenylamid

Purity: 98+% CAS No.: 93-98-1 Molecular Formula: C6H5NHCOC6H5 Formula Weight: 197.24 Odour: Odourless Melting Point: 162-166° Boiling Point: 117°/10mm Beilstein Reference: 1102980 Merck Reference: 14,1061 MDL No.: MFCD00003069

Formel: C₁₃H₁₁NO
Kogepunkt: 117 °C (10 mmHg)
Smeltepunkt: 162…166 °C
Massefylde: 1,32
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003069
CAS-nummer: 93-98-1
EINECS: 202-292-7
Merck Index: 14,1061

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR