Benzoylchlorid 98+% ACS

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Benzoic acid chloride

402030025 402030050 402035000
ACRO402030025EA 1890 DKK
ACRO402030025 ACRO402030050 ACRO402035000
Benzoylchlorid 98+% ACS
Benzoylchlorid
Formel: C₇H₅ClO
Kogepunkt: 197,2 °C (1013 hPa)
Smeltepunkt: –1 °C
Massefylde: 1,21 g/cm³
Flammepunkt: 93 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000653
CAS-nummer: 98-88-4
EINECS: 202-710-8
UN nummer: 1736
ADR: 8,II
Merck Index: 13,01113

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5540 (20°C, 589 nm)
Residue after ignition ≤0.005 %
Titration excess NaOH 98.0 to 100.5 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Phosphorous compounds (as P) ≤0.002 %
Freezing point -2.0 to 0.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR