4-Cyanobenzoylchlorid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

154550050 154550250
ACRO154550050EA 431 DKK
ACRO154550050 ACRO154550250
4-Cyanobenzoylchlorid 98%
4-Cyanobenzoylchlorid
Formel: C₈H₄ClNO
Kogepunkt: 150…151 °C (25 mmHg)
Smeltepunkt: 66…70 °C
MDL Number: MFCD00001822
CAS-nummer: 6068-72-0
EINECS: 228-005-5
UN nummer: 1759
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 66 to 70 °C
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR