2,4,6-Trimethylphenylisocyanat ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
L12611.06
L12611.03
L12611.06EA
782
DKK
L12611.06
L12611.03
2,4,6-Trimethylphenylisocyanat ≥98%
2,4,6-Trimethylphenylisocyanat
Purity: 98%
CAS No.: 2958-62-5
Molecular Formula: C10H11NO
Formula Weight: 161.21
Storage Temperature: 2°C to 8°C
Flash Point: 104°(219°F)
Odour: Irritating
Melting Point: 43-46°
Boiling Point: 218-220°
Beilstein Reference: 2085601
UN No.: 2811
MDL No.: MFCD00019909
Formel:
C₁₀H₁₁NO Kogepunkt: 218…220 °C Smeltepunkt: 43…46 °C Flammepunkt: 104 °C (219 °F) |
MDL Number:
MFCD00019909 CAS-nummer: 2958-62-5 UN nummer: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...