Powder funnels, stainless steel
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A716-300 A716-100 A716-200
SAMPA716-300EA 4700 DKK
SAMPA716-300 SAMPA716-100 SAMPA716-200
Powder funnels, stainless steel
Tragte Pulvertragte
These powder funnels are made from hygienic 316 stainless steel.
  • Satin finish
  • 2" tri ferrule fitting/opening at spout base
  • Very good chemical and temperature resistance
The powder funnel has a wide 50 mm diameter outlet to help prevent blackages. The outlet has a 2" ferrule so the the powder funnel can be clamped to a suitable branch.