Jars, round, wide mouth, PET with thread
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9-4000-0500-93 9-4000-0500-82
RIXI9-4000-0500-93EA 3270 DKK
RIXI9-4000-0500-93 RIXI9-4000-0500-82
Jars, round, wide mouth, PET with thread
Prøvecontainere Allround kasser
Made of PET, clear.
  • Good chemical resistance
  • Lightweight
  • Wide neck for easy filling and cleaning
Ideal for collecting, storing and transporting samples.
Bestillingsoplysninger: Screw lids must be ordered seperately.
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904