2,4,6-Trichlorbenzonitril ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A14609.03 A14609.06 A14609.14
A14609.03EA 251 DKK
A14609.03 A14609.06 A14609.14
2,4,6-Trichlorbenzonitril ≥97%
2,4,6-Trichlorbenzonitril

Purity: 97+% CAS No.: 6575-05-9 Molecular Formula: C7H2Cl3N Formula Weight: 206.46 Melting Point: 80-82° UN No.: 3439 MDL No.: MFCD00052711

Formel: C₇H₂Cl₃N
Molekylvægt: 206.46 g/mol
Smeltepunkt: 80…82 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052711
CAS-nummer: 6575-05-9
UN nummer: 3439
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR