3-Cyanobenzaldehyd 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Formylbenzonitrile
163900050
163900250
163900010
ACRO163900050EA
1010
DKK
ACRO163900050
ACRO163900250
ACRO163900010
3-Cyanobenzaldehyd 99%
3-Cyanobenzaldehyd
Formel:
C₈H₅NO Kogepunkt: 210 °C (1013 hPa) Smeltepunkt: 79…81 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003344 CAS-nummer: 24964-64-5 EINECS: 246-549-1 UN nummer: 3439 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to light yellow Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 76 to 80 °C |
GC | ≥98.5 % |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...