(2-Bromethoxy)benzen ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 589-10-6
Molecular Formula: C6H5O(CH2)2Br
Formula Weight: 201.07
Flash Point: 65°(149°F)
Melting Point: 31-35°
Boiling Point: 135-137°/20mm
Beilstein Reference: 508290
MDL No.: MFCD00000234
Formel:
BrCH₂CH₂OC₆H₅ Kogepunkt: 135…137 °C (20 mmHg) Smeltepunkt: 31…35 °C Massefylde: 1,35 Flammepunkt: 65 °C (149 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000234 CAS-nummer: 589-10-6 EINECS: 209-634-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...