4-Brom-o-anisidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Bromo-2-methoxyaniline, 2-Amino-5-bromoanisole, 2-Methoxy-4-bromoaniline

A2027.0005 A2027-1G A2027-25G A2027-5G
TCIAA2027.0005EA 0 DKK
TCIAA2027.0005 TCIAA2027-1G TCIAA2027-25G TCIAA2027-5G
4-Brom-o-anisidin ≥98.0% (by GC, titration analysis)
4-Brom-o-anisidin
Formel: C₇H₈BrNO
Molekylvægt: 202.05 g/mol
MDL Number: MFCD01204266
CAS-nummer: 59557-91-4
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c0811bfc60ee31f5a89cee9f68d31b21f3fe0e&vid=f6c0811bfc60ee31f5a89cee9f68d31b21f3fe0e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR