3-Brom-N,N-dimethylanilin ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 16518-62-0
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Refractive Index: 1.6015
Melting Point: 11°
Boiling Point: 126°/14mm
Beilstein Reference: 2079908
MDL No.: MFCD00045020
Formel:
C₈H₁₀BrN Kogepunkt: 126 °C (14 mmHg) Smeltepunkt: 11 °C Massefylde: 1,365 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00045020 CAS-nummer: 16518-62-0 |