2,3-Dimethoxyphenylboronsyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,3-Dimethoxybenzeneboronic acid, (2,3-Dimethoxyphenyl)metaboric acid, Dihydroxy(2,3-dimethoxyphenyl)borane

B24125.06 B24125.14
B24125.06EA 1160 DKK
B24125.06 B24125.14
2,3-Dimethoxyphenylboronsyre ≥98%
2,3-Dimethoxyphenylboronsyre

Purity: 98% CAS No.: 40972-86-9 Molecular Formula: C8H11BO4 Formula Weight: 181.98 Melting Point: 67-71° Beilstein Reference: 3051295

Formel: C₈H₁₁BO₄
Smeltepunkt: 67…71 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02683112
CAS-nummer: 40972-86-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR