Piperonylsyre 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4-(Methylenedioxy)benzoic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, 1,3-Benzodioxole-5-carboxylic acid

131370250 131371000 131370050
ACRO131370250EA 602 DKK
ACRO131370250 ACRO131371000 ACRO131370050
Piperonylsyre 99%
Piperonylsyre
Formel: C₈H₆O₄
Kogepunkt: 180 °C (1 -rr)
Smeltepunkt: 229…235 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005830
CAS-nummer: 94-53-1
EINECS: 202-342-8
Merck Index: 12,07630

Bestil nu

Specifikationer Testresultater

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Melting point 229 to 235 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR