5-Methylisatin ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5-Methylindoline-2,3-dione, 5-Methyl-1H-indole-2,3-dione
L08009.03
L08009.06
L08009.14
L08009.03EA
445
DKK
L08009.03
L08009.06
L08009.14
5-Methylisatin ≥97%
5-Methylisatin
Purity: 97%
CAS No.: 608-05-9
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: ca 180° dec.
Beilstein Reference: 123738
Formel:
C₉H₇NO₂ Molekylvægt: 161.16 g/mol Smeltepunkt: ∼ 180 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00005721 CAS-nummer: 608-05-9 EINECS: 210-152-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...