1-Methylpyrrolidin for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-Methylpyrrolidine

M-Clarity™ quality level = MQ200

8.16146.0010 8.16146.0100
8.16146.0010EA 262 DKK
8.16146.0010 8.16146.0100
1-Methylpyrrolidin for synthesis, Sigma-Aldrich®
1-Methylpyrrolidin
Formel: C₅H₁₁N
Kogepunkt: 80 °C (1013 hPa)
Smeltepunkt: –76 °C
Massefylde: 0,799 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003173
CAS-nummer: 120-94-5
EINECS: 204-438-5
UN nummer: 3286
ADR: 3 (6.1, 8),II

Bestil nu

Specifikationer Testresultater

Solubility 213 g/l
Molar Mass 85.15 g/mol
Boiling Point 81 °C
Vapor Pressure 100 hPa (20 °C)
Flash Point -3 °C
Explosion Limit 1.1 - 9.1 %(V)
Density 0.799 g/cm3
pH 11.3 (1 g/l, H2O)
Ignition Point 170 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR