1-Methylpyrrolidin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
N-Methylpyrrolidine
B23799.22
B23799.36
B23799.22EA
415
DKK
B23799.22
B23799.36
1-Methylpyrrolidin ≥98%
1-Methylpyrrolidin
Purity: 98%
CAS No.: 120-94-5
Molecular Formula: C5H11N
Formula Weight: 85.15
Flash Point: -18°(0°F)
Refractive Index: 1.4247
Odour: Amine-like
pH: 11.3
Melting Point: -90°
Boiling Point: 80-81°
UN No.: 3286
MDL No.: MFCD00003173
Formel:
C₅H₁₁N Kogepunkt: 80…81 °C Smeltepunkt: –90 °C Massefylde: 0,799 Flammepunkt: –18 °C (0 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003173 CAS-nummer: 120-94-5 EINECS: 204-438-5 UN nummer: 3286 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...