N-Methyl-2-pyrrolidon (NMP), EMPLURA®, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone
MQ200
8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
8.06072.1000EA 534 DKK
8.06072.1000 8.06072.2500 8.06072.9025 8.06072.9210 8.06072.9011 8.06072.9180
N-Methyl-2-pyrrolidon (NMP), EMPLURA®, Supelco®
N-Methyl-2-pyrrolidon (NMP)
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Formel: C₅H₉NO
Molekylvægt: 99.13 g/mol
Kogepunkt: 202
Smeltepunkt: –24 °C
Massefylde: 1.033 g/cm³ (20 °C)
Flammepunkt: 91
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003193
CAS-nummer: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM