3-Butyn-1-ol 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

157330050 157330250 157331000
ACRO157330050EA 367 DKK
ACRO157330050 ACRO157330250 ACRO157331000
3-Butyn-1-ol 97%
3-Butyn-1-ol
Formel: C₄H₆O
Kogepunkt: 128,8 °C (1013 hPa)
Massefylde: 0,926 g/cm³
Flammepunkt: 36 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002955
CAS-nummer: 927-74-2
EINECS: 213-161-9
UN nummer: 1986
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -10° to -20° (20°C, 589 nm) (c=4 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR