4-Pentynoic acid 95%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 98%
CAS No.: 6089-09-4
Molecular Formula: C5H6O2
Formula Weight: 98.10
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Melting Point: 51-58°
Boiling Point: 109-110°/30mm
Beilstein Reference: 1742047
UN No.: 3261
MDL No.: MFCD00004407
Formel:
C₅H₆O₂ Kogepunkt: 109…110 °C (30 mmHg) Smeltepunkt: 51…58 °C Flammepunkt: 75 °C (167 °F) |
MDL Number:
MFCD00004407 CAS-nummer: 6089-09-4 EINECS: 228-028-0 UN nummer: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...