3-Butyn-2-on 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
But-3-yn-2-one
330440025
330440100
ACRO330440025EA
1040
DKK
ACRO330440025
ACRO330440100
3-Butyn-2-on 96%
3-Butyn-2-on
Formel:
C₄H₄O Kogepunkt: 85 °C (760 mmHg) Massefylde: 0,87 g/cm³ Flammepunkt: –1 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00008775 CAS-nummer: 1423-60-5 EINECS: 215-834-2 UN nummer: 1992 ADR: 3,II |
Specifikationer Testresultater
Appearance | Clear yellow to orange-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4045 to 1.4095 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...