5-Cyano-1-pentyn ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L02604.03 L02604.06
L02604.03EA 387 DKK
L02604.03 L02604.06
5-Cyano-1-pentyn ≥98%
5-Cyano-1-pentyn

Purity: 98% CAS No.: 14918-21-9 Molecular Formula: HC≡CCH2CH2CH2CN Formula Weight: 93.13 Storage Temperature: 2°C to 8°C Flash Point: 42°(107°F) Refractive Index: 1.4410 Boiling Point: 115-117° Beilstein Reference: 1735926 UN No.: 1992 MDL No.: MFCD00001978

Formel: C₆H₇N
Kogepunkt: 115…117 °C
Massefylde: 0,889
Flammepunkt: 42 °C (107 °F)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001978
CAS-nummer: 14918-21-9
UN nummer: 1992
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR