Sinapinsyre, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

M-Clarity™ quality level = MQ100

85429-1G 85429-5G
85429-1GEA 589 DKK
85429-1G 85429-5G
Sinapinsyre, Supelco®
Sinapinsyre
Formel: C₁₁H₁₂O₅
Kogepunkt: 378 °C (1013 hPa)
Smeltepunkt: 200…202 °C
MDL Number: MFCD00004401
CAS-nummer: 530-59-6
EINECS: 208-487-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=928488ef3f77c025eddb437e8011266bf6bab0da&vid=6337ff25b90d8ce60e2d1c1d65bd8a069d65c1c5&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR