3-Chlorkanelsyre predominantly trans ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(3-Chlorophenyl)acrylic acid, (2E)-3-(3-Chlorophenyl)acrylic acid
A13416.09
A13416.18
A13416.30
A13416.09EA
375
DKK
A13416.09
A13416.18
A13416.30
3-Chlorkanelsyre predominantly trans ≥98%
3-Chlorkanelsyre
Purity: 98+%
CAS No.: 1866-38-2
Molecular Formula: C9H7ClO2
Formula Weight: 182.61
Melting Point: 161-164°
Beilstein Reference: 2207378
Formel:
C₉H₇ClO₂ Smeltepunkt: 161…164 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004384 CAS-nummer: 1866-38-2 EINECS: 217-478-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...