cis-2-Penten-1-ol ≥97% remainder mainly trans-isomer
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 97%
CAS No.: 1576-95-0
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: 48°(118°F)
Refractive Index: 1.4360
Boiling Point: 138°
Beilstein Reference: 1719473
UN No.: 1987
MDL No.: MFCD00063208
Formel:
C₅H₁₀O Kogepunkt: 138 °C Massefylde: 0,853 Flammepunkt: 48 °C (118 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00063208 CAS-nummer: 1576-95-0 EINECS: 216-415-7 UN nummer: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...