(4-fluorphenyl)eddikesyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Fluorophenyl)acetic acid

A10646.06 A10646.14 A10646.22
A10646.06EA 276 DKK
A10646.06 A10646.14 A10646.22
(4-fluorphenyl)eddikesyre ≥98%
(4-fluorphenyl)eddikesyre

Purity: 98% CAS No.: 405-50-5 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 100°(212°F) Odour: Odourless Boiling Point: 164°/2mm Beilstein Reference: 972145 Merck Reference: 14,4177 MDL No.: MFCD00004343

Formel: (C₆H₄F)CH₂COOH
Kogepunkt: 164 °C (2 mmHg)
Smeltepunkt: 81…85 °C
Flammepunkt: >100 °C (212 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004343
CAS-nummer: 405-50-5
EINECS: 206-972-4
Merck Index: 14,4177

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR