(3,4,5-Trimethoxyphenyl)eddikesyre 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,4,5-Trimethoxyphenylacetic acid

139990100 139990050 139990500
ACRO139990100EA 529 DKK
ACRO139990100 ACRO139990050 ACRO139990500
(3,4,5-Trimethoxyphenyl)eddikesyre 99%
(3,4,5-Trimethoxyphenyl)eddikesyre
Formel: (C₆H₂(OCH₃)₃)CH₂COOH
Kogepunkt: 352 °C (1013 hPa)
Smeltepunkt: 117…120 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004336
CAS-nummer: 951-82-6
EINECS: 213-456-2

Bestil nu

Specifikationer Testresultater

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 120 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR