(4-Chlorphenyl)eddikesyre 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

160721000 160725000 160720050 160720025
ACRO160721000EA 604 DKK
ACRO160721000 ACRO160725000 ACRO160720050 ACRO160720025
(4-Chlorphenyl)eddikesyre 98%
(4-Chlorphenyl)eddikesyre
Formel: (C₆H₄Cl)CH₂COOH
Kogepunkt: 292 °C (1013 hPa)
Smeltepunkt: 103…107 °C
Massefylde: 1,43 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004344
CAS-nummer: 1878-66-6
EINECS: 217-521-6

Bestil nu

Specifikationer Testresultater

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
Water ≤0.2 % (K.F.)
HPLC ≥97.5 %
Appearance of solution (5% soln. in CHCl3) clear
Titration with NaOH 97.5 to 101.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR