Isobutanol, EMPLURA®, Supelco®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Methyl-1-propanol
M-Clarity™ quality level = MQ200
1.00985.6025
1.00985.9190
1.00985.6025EA
7210
DKK
1.00985.6025
1.00985.9190
Isobutanol, EMPLURA®, Supelco®
Isobutanol
Formel:
(CH₃)₂CHCH₂OH Kogepunkt: 107,9 °C (1013 hPa) Smeltepunkt: –108 °C Massefylde: 0,802 g/cm³ (20 °C) Flammepunkt: 28 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004740 CAS-nummer: 78-83-1 EINECS: 201-148-0 UN nummer: 1212 ADR: 3,III Merck Index: 13,05148 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...