2-Ethylbutanol ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B21026.0F
B21026.AE
B21026.AP
B21026.0FEA
17990
DKK
B21026.0F
B21026.AE
B21026.AP
2-Ethylbutanol ≥99%
2-Ethylbutanol
Purity: 99%
CAS No.: 97-95-0
Molecular Formula: (CH3CH2)2CHCH2OH
Formula Weight: 102.18
Flash Point: 58°(136°F)
Refractive Index: 1.4220
Melting Point: -52°
Boiling Point: 146-148°
Beilstein Reference: 1731254
UN No.: 1987
MDL No.: MFCD00004744
Formel:
C₆H₁₄O Kogepunkt: 146…148 °C Smeltepunkt: –52 °C Massefylde: 0,831 Flammepunkt: 58 °C (136 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004744 CAS-nummer: 97-95-0 EINECS: 202-621-4 UN nummer: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...