3,3-Dimethylglutarsyre ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid

B20388.09 B20388.18 B20388.30
B20388.09EA 384 DKK
B20388.09 B20388.18 B20388.30
3,3-Dimethylglutarsyre ≥98%
3,3-Dimethylglutarsyre

Purity: 98+% CAS No.: 4839-46-7 Molecular Formula: C7H12O4 Formula Weight: 160.17 Melting Point: 98-103° Boiling Point: 89-90°/2mm MDL No.: MFCD00002716

Formel: C₇H₁₂O₄
Molekylvægt: 160.17 g/mol
Kogepunkt: 89…90 °C (2 mmHg)
Smeltepunkt: 98…103 °C
Massefylde: 1.42
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002716
CAS-nummer: 4839-46-7
EINECS: 225-425-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR