L(-)-Glutathion (oxidized form)

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)

23130.01 23130.02 23130.03
SERA23130.01EA 253.59 DKK
SERA23130.01 SERA23130.02 SERA23130.03
L(-)-Glutathion (oxidized form)
L(-)-Glutathion (oxidized form)
Formel: C₂₀H₃₂N₆O₁₂S₂
Molekylvægt: 612.64 g/mol
Smeltepunkt: 185
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00063106
CAS-nummer: 27025-41-8
EINECS: 248-170-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR