VWR®, Shoe Covers, Anti-Slip
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-9081EA 726 DKK
113-9081 113-9080 113-9079 113-9082
VWR®, Shoe Covers, Anti-Slip
Engangsbeklædning
Disposable shoe covers made of CPE film.
  • Free from natural rubber latex
  • Anti-slip performance
L×W: 410×170 mm
Weight: 10, 13 or 16 g/pcs
* One size fits all
Certificeringer: Cat. I