Safety ankle boots, lace-up, Basic, Alex, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16521/36 16521/37 16521/38 16521/39 16521/40 16521/41 16521/42 16521/43 16521/44 16521/45 16521/46 16521/47 16521/48
ELTN16521/36EA 462 DKK
ELTN16521/36 ELTN16521/37 ELTN16521/38 ELTN16521/39 ELTN16521/40 ELTN16521/41 ELTN16521/42 ELTN16521/43 ELTN16521/44 ELTN16521/45 ELTN16521/46 ELTN16521/47 ELTN16521/48
Safety ankle boots, lace-up, Basic, Alex, Jori
Sko og støvler
Durable safety ankle boots made from cowhide leather and hydrophobised CORDURA® (tear and abrasion resistant, dirt and water repellent), reflective materials, skin friendly textile lining, a full padded bellows tongue and collar padding plus a dual-density PU/PU New Basic sole (oil, fuel and abrasion resistant, also heat resistant up to approx. 130 °C).
  • Climate-regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full length inlay sole
  • Anti-static soft fleece insole
  • PU scuff cap protects the upper material
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/blue
Certificeringer: EN ISO 20345; S3, SRC