Safety ankle boots, lace-up, jo_Max Mid, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
16731/38 16731/39 16731/40 16731/41 16731/42 16731/43 16731/44 16731/45 16731/46 16731/47 16731/48
ELTN16731/38EA 529 DKK
ELTN16731/38 ELTN16731/39 ELTN16731/40 ELTN16731/41 ELTN16731/42 ELTN16731/43 ELTN16731/44 ELTN16731/45 ELTN16731/46 ELTN16731/47 ELTN16731/48
Safety ankle boots, lace-up, jo_Max Mid, Jori
Sko og støvler
Antistatic safety ankle boot made from hydrophobised nubuck leather, mesh inserts and reflective materials, skin-friendly textile lining, a full, padded bellows tongue and collar, and a dual-density rubber/PU jo_MAX sole (3,0 mm profile, oil, fuel and chemical resistant, temperature resistant from approx. -20 and +200 °C).
  • Climate regulating, breathable textile lining
  • Heel pocket lining made from sturdy abrasion resistant microfibre lining
  • Full-length inlay sole
  • Protective composite toecap (200 J)
  • Metal-free, puncture resistant midsole
Ideal for useMore Product Information