Octan-1-sulfonsyre natrium salt for HPLC
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Sodium octane-1-sulphonate
384770250 384771000 384770050 384775000
ACRO384770250EA 3880 DKK
ACRO384770250 ACRO384771000 ACRO384770050 ACRO384775000
Octan-1-sulfonsyre natrium salt for HPLC
Octan-1-sulfonsyre natrium salt
Formel: C₈H₁₇NaO₃S
Molekylvægt: 216.28 g/mol
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007544
CAS-nummer: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤2 %
UV at 240 nm A: ≤0.05
Titration after Ion exchange ≥98.0 %