Ethyl methansulphonat 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
205260100
205260250
205261000
205265000
ACRO205260100EA
874
DKK
ACRO205260100
ACRO205260250
ACRO205261000
ACRO205265000
Ethyl methansulphonat 99%
Ethyl methansulphonat
Formel:
C₃H₈O₃S Molekylvægt: 124.16 g/mol Kogepunkt: 213…214 °C (760 mmHg) Massefylde: 1.16 g/cm³ Flammepunkt: 100 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007559 CAS-nummer: 62-50-0 EINECS: 200-536-7 ADR: 6.1 II Merck Index: 13,03859 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4190 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...