1-Tetradecanthiol ≥94%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: n-Tetradecyl mercaptan, 1-Mercaptotetradecane, n-Tetradecanethiol, N-tetradecylmercaptan

L09401.06 L09401.14
L09401.06EA 400 DKK
L09401.06 L09401.14
1-Tetradecanthiol ≥94%
1-Tetradecanthiol

Purity: 94% CAS No.: 2079-95-0 Molecular Formula: C14H30S Formula Weight: 230.46 Flash Point: 110°(230°F) Refractive Index: 1.4610 Odour: Unpleasant Melting Point: 6-7° Boiling Point: 176-180°/22mm Beilstein Reference: 1742653 MDL No.: MFCD00022099

Formel: C₁₄H₃₀S
Kogepunkt: 176…180 °C (22 mmHg)
Smeltepunkt: 6…7 °C
Massefylde: 0,850
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00022099
CAS-nummer: 2079-95-0
EINECS: 218-209-2
UN nummer: 3334
ADR: 9,

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR