Diethylsulfat 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
114560010
114560025
114560250
114560100
ACRO114560010EA
848
DKK
ACRO114560010
ACRO114560025
ACRO114560250
ACRO114560100
Diethylsulfat 99%
Diethylsulfat
Formel:
C₄H₁₀O₄S Kogepunkt: 208 °C (760 mmHg) Smeltepunkt: –24 °C Massefylde: 1,1795 g/cm³ Flammepunkt: 104 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00009099 CAS-nummer: 64-67-5 EINECS: 200-589-6 UN nummer: 1594 ADR: 6.1,II Merck Index: 13,03156 |
Specifikationer Testresultater
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3990 to 1.4010 (20°C, 589 nm) |
Water | ≤0.15 % |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...