Smørsyreanhydrid 98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
108120025 108121000 108125000
ACRO108120025EA 1220 DKK
ACRO108120025 ACRO108121000 ACRO108125000
Smørsyreanhydrid 98%
Smørsyreanhydrid
Formel: C₈H₁₄O₃
Molekylvægt: 158.2 g/mol
Kogepunkt: 195 °C (760 mmHg)
Smeltepunkt: –73 °C
Massefylde: 0.963 g/cm³
Flammepunkt: 82 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009389
CAS-nummer: 106-31-0
EINECS: 203-383-4
UN nummer: 2739
ADR: 8 III
Merck Index: 13,01594
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
GC ≥97.5 %
UV at 260 nm A: ≤0.15
Appearance of solution faint yellow
NMR (1H-NMR) Chondroitin sulfate (over-sulfated): Absent
Protein ≤0.5 % (dried substance)
pH 5.5 to 8.0 (0.1g in 10ml water)