2-Nitro-1-propanol ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Hydroxy-2-nitropropane

43843.14 43843.06
43843.14EA 2330 DKK
43843.14 43843.06
2-Nitro-1-propanol ≥98%
2-Nitro-1-propanol

Purity: 98% CAS No.: 2902-96-7 Molecular Formula: CH3CH(NO2)CH2OH Formula Weight: 105.09 Flash Point: 100°(212°F) Physical Form: Liquid Density: 1.185 Refractive Index: 1.4390 Boiling Point: 73°/1mm MDL No.: MFCD00007398

Formel: C₃H₇NO₃
Molekylvægt: 105.09 g/mol
Kogepunkt: 73 °C (1 mmHg)
Massefylde: 1.185
Flammepunkt: 100 °C (212 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007398
CAS-nummer: 2902-96-7
EINECS: 220-797-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR