VWR® Basic, Shoe Covers, Anti-Skid
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-8285EA 1450 DKK
113-8285 113-8287
VWR® Basic, Shoe Covers, Anti-Skid
Engangsbeklædning
These disposable shoe covers are made from non woven, laminated, PP fabric.
  • Anti-skid sole
  • Free from natural rubber latex
  • Available in white and blue
L×H: 410×170 mm
Weight: 60 g/m²
* One size fits all
Emballering: 10 pieces per bundle, 10 bundles per bag, 3 bags per case.