Aminomethansulfonsyre 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B23441.14 B23441.22 B23441.36
B23441.14EA 288 DKK
B23441.14 B23441.22 B23441.36
Aminomethansulfonsyre 97%
Aminomethansulfonsyre

Purity: 97% CAS No.: 13881-91-9 Molecular Formula: H2NCH2SO3H Formula Weight: 111.12 pH: 3.2 Melting Point: ca 180° dec. Beilstein Reference: 1811756 UN No.: 2585 MDL No.: MFCD00008124

Formel: CH₅NO₃S
Smeltepunkt: ∼ 180 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008124
CAS-nummer: 13881-91-9
EINECS: 237-649-6
ADR: 8,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR